Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals NP-40 lysis buffer

Catalog No. p-7041936
Click to view available options
Quantity:
250 mL
500 mL

NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

Solubility
It is soluble in water.

Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Quantity 500 mL
Color Colorless
Content And Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.