Learn More
Thermo Scientific™ NP-40 Surfact-Amps™ Detergent Solution, 50 mL PROMO
Description
Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.
This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.
Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
NP-40 – a nonionic detergent for use in various protein methods
Accurate – precise 10% detergent solution in ultrapure water
Easy-to-use – solution is simple to dispense and dilute for use
Exceptionally pure – less than 1.0μeq/mL peroxides and carbonyls
Stable – packed in glass ampules under inert nitrogen gas
Detergent Properties:
- Molecular Weight: 617
- Detergent Class: Nonionic
- Aggregation Number: 149
- Micelle Molecular Weight: 90,000
- Critical Micelle Concentration (CMC): 0.29mM (0.0179%, w/v)
- Cloud Point: 80°C
- Dialyzable: No
;
Specifications
Specifications
| Product Type | Detergent |
| Content And Storage | Store between 20°C and 25°C |
| Form | Liquid |
| Product Line | Surfact-Amps |
| Reagent Type | Detergent Solution |
| Quantity | 50 mL |
| Format | HDPE Bottle |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.