Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPAS1 Monoclonal Antibody (OTI1C9)
GREENER_CHOICE

Catalog No. PIMA527189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527189 Supplier Invitrogen™ Supplier No. MA527189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPAS1 Monoclonal Antibody for Western Blot, IHC (P)

NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NPAS1
Gene Accession No. Q99742
Gene Alias basic-helix-loop-helix-PAS protein MOP5; BHLHE11; Class E basic helix-loop-helix protein 11; member of PAS protein 5; member of PAS superfamily 5; MOP5; neuronal PAS domain protein 1; neuronal PAS domain-containing protein 1; Neuronal PAS1; Npas1; PAS domain-containing protein 5; PASD5
Gene Symbols NPAS1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 319-522 of NPAS1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4861
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.