Learn More
NPAS1 Mouse anti-Human, Clone: OTI1H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809585

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.Specifications
NPAS1 | |
Monoclonal | |
Unconjugated | |
NPAS1 | |
MOP5, PASD5, bHLHe11 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4861 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99742 | |
NPAS1 | |
Human recombit protein fragment corresponding to amino acids 319-522 of human NPAS1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.