Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPAS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595855
Description
Immunogen sequence: KTTVLEKVIG FLQKHNEVSA QTEICDIQQD WKPSFLSNEE FTQLMLEALD GFIIAVTTDG SIIYVSDSIT PLLGHLPSDV MDQNLLNFLP EQEHSEVYKI LSSHMLVTDS PSPEYLKSDS DLEFYCHLLR GSLNPKEFPT YEYIKFVGNF RSYNNVPSPS CNGFDNTLSR PCRVPLGKEV CFIATVRLAT PQFLKE.
Members of the basic helix-loop-helix-PER-ARNT-SIM (bHLH-PAS) family are transcription factors that contain a bHLH DNA binding domain located amino-terminal to a PAS domain. Neuronal PAS domain protein 2 (NPAS2, also designated PAS 4/MOP4) is a member of the bHLH-PAS family and the PAS superfamily. NPAS2, which maps to chromosome 2p11.2-2q13, is expressed primarily in the neurons during the first week of postnatal development. The pattern of NPAS2 expression temporally matches the development of learning and memory, and spatially matches the frontal association/limbic forebrain pathway. NPAS2 may serve a regulatory role in the development and maintenance of long-term memory, and may be required for the processing of complex sensory information. NPAS2 and MOP3 form a transcriptionally active heterodimer which binds to a CACGTGA-containing DNA element and drives transcription from a linked luciferase reporter gene.
Specifications
NPAS2 | |
Polyclonal | |
Unconjugated | |
Npas2 | |
Basic-helix-loop-helix-PAS protein MOP4; bHLHe9; Class E basic helix-loop-helix protein 9; member of PAS protein 4; member of PAS superfamily 4; MOP4; neuronal PAS domain protein 2; neuronal PAS domain-containing protein 2; Neuronal PAS2; NPAS2; PAS domain-containing protein 4; PASD4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
316351, 4862 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99743 | |
Npas2 | |
A synthetic peptide corresponding to a sequence within amino acids 45-240 of human NPAS2 (NP_0025092). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction