Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPAS4 Monoclonal Antibody (N408/79), FITC
GREENER_CHOICE

Catalog No. PIMA545750
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545750 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545750 Supplier Invitrogen™ Supplier No. MA545750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45750 was sufficient for detection of NPAS4 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa.approximately This antibody was formerly sold as clone S408-79.

NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N408/79
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene NPAS4
Gene Accession No. Q8BGD7, Q8CJH6, Q8IUM7
Gene Alias bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS
Gene Symbols NPAS4
Host Species Mouse
Immunogen Fusion protein amino acids 597-802 (C-terminus) of rat Npas4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225872, 266734, 266743
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.