Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Npas4 Mouse anti-Rat, Alkaline Phosphatase, Clone: S408-79, Abnova™

Catalog No. 89338697 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-697 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-697 Supplier Abnova Corporation Supplier No. MAB19193

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody.

Npas4 Monoclonal antibody specifically detects Npas4 in Rat samples. It is validated for ICC, WB-Ti, IHC-P, IF.

Specifications

Antigen Npas4
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S408-79
Conjugate Alkaline Phosphatase (AP)
Description neuronal PAS domain protein 4
Gene Npas4
Gene Alias Nxf
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 597-802 at C-terminus of rat Npas4
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 266734
Target Species Rat
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.