Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPAS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142642
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142642 Supplier Invitrogen™ Supplier No. PA5142642
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.

NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NPAS4
Gene Accession No. Q8BGD7, Q8CJH6, Q8IUM7
Gene Alias bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS
Gene Symbols NPAS4
Host Species Goat
Immunogen Peptide with sequence CRFNTSKSLRRQS (aa 149-161).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225872, 266734, 266743
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.