Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565419 Supplier Invitrogen™ Supplier No. PA565419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNQAVSPNFSQ GSAIIIASPV QPVLQGMVGM IPVSVVGQNG NNFSTPPRQV LHMPLTAPVC NRSIPQFPVP PKSQKAQGLR NKPCIGKQVN NLVDSSGHSV Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.

Required for progression through the G1 and S phases of the cell cycle and for S phase entry. Activates transcription of the histone H2A, histone H2B, histone H3 and histone H4 genes in conjunction with MIZF. Also positively regulates the ATM, MIZF and PRKDC promoters. Transcriptional activation may be accomplished at least in part by the recruitment of the NuA4 histone acetyltransferase (HAT) complex to target gene promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAT
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPAT
Gene Accession No. Q14207
Gene Alias CAND3; E14; E14/NPAT; NPAT; Nuclear protein of the ataxia telangiectasia mutated locus; nuclear protein of the ATM locus; nuclear protein, ataxia-telangiectasia locus; nuclear protein, coactivator of histone transcription; nuclear protein, co-activator of histone transcription; p220; Protein NPAT
Gene Symbols NPAT
Host Species Rabbit
Immunogen Recombinant Human NPAT. Recombinant protein control fragment (Product #RP-106102).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.