Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPB Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NPB |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4943
![]() ![]() |
Proteintech Group Inc
255171AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Neuropeptide B is a endrogenous ligand of the orphan G protein coupled receptor GPR7. NPB and GPR7 exist in close proximity in the rat hypothalamus, and are ideally positioned to modulate neuroendocrine functions. NPB-immunoreactivity was enriched in many regions within the hypothalamus which also contained high levels of GPR7 mRNA including the ventrormedial hypothalamic nucleus, dorsomedial hypothalamic nucleus, arcuate nucleus, supraoptic retrochiasmatic nucleus, and in the area ventral to the zona incerta.Specifications
NPB | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8NG41 | |
256933 | |
NPB Fusion Protein Ag21877 | |
Primary | |
-20°C | |
NPB |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hL7 Neuropeptide B 29, hL7C, hPPL7, neuropeptide B, NPB, PPL7, PPNPB, Preproprotein L7 | |
NPB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title