Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPBWR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100946 Supplier Invitrogen™ Supplier No. PA5100946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR7.

Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals. Has a higher affinity for neuropeptide B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPBWR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NPBWR1
Gene Accession No. P48145, P49681
Gene Alias G protein-coupled receptor 7; Gpr7; G-protein coupled receptor 7; MGC129755; Nbpwr1; neuropeptide B/W receptor 1; neuropeptides B/W receptor 1; neuropeptides B/W receptor type 1; NPBWR1; opioid-somatostatin-like receptor 7
Gene Symbols NPBWR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPBWR1(Accession P48145), corresponding to amino acid residues S289-A328.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226304, 2831
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.