Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPBWR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533977
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533977 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533977 Supplier Invitrogen™ Supplier No. PA533977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Elephant, Panda (100%) Marmoset, Mouse, Rat, Bovine, Horse (95%) Rabbit (84%).

Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals. Has a higher affinity for neuropeptide B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPBWR1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NPBWR1
Gene Accession No. P48145, Q56UD9
Gene Alias G protein-coupled receptor 7; Gpr7; G-protein coupled receptor 7; MGC129755; Nbpwr1; neuropeptide B/W receptor 1; neuropeptides B/W receptor 1; neuropeptides B/W receptor type 1; NPBWR1; opioid-somatostatin-like receptor 7
Gene Symbols NPBWR1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 2nd extracellular domain of human NPBWR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2831, 297795
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.