Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPBWR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533979
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533979 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533979 Supplier Invitrogen™ Supplier No. PA533979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey, Marmoset (94%) Gibbon (88%).

The protein encoded by this gene is an integral membrane protein and G protein-coupled receptor. The encoded protein is similar in sequence to another G protein-coupled receptor (GPR7), and it is structurally similar to opioid and somatostatin receptors. This protein binds neuropeptides B and W. This gene is intronless and is expressed primarily in the frontal cortex of the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPBWR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NPBWR2
Gene Accession No. P48146
Gene Alias G protein-coupled receptor 8; GPR8; G-protein coupled receptor 8; neuropeptides B/W receptor 2; neuropeptides B/W receptor type 2; NPBWR2; opioid-somatostatin-like receptor 8
Gene Symbols NPBWR2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from N-terminal extracellular domain of human NPBWR2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.