Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599027
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599027 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599027 Supplier Invitrogen™ Supplier No. PA599027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPC1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mutations in the Niemann-Pick disease type C1 (NPC1) gene result in a fatal progressive neurodegenerative disorder characterized by an abnormal sequestration of lipids including cholesterol and glycosphingolipids. The NPC1 protein is a large protein that resides in the limiting membrane of endosomes and lysosomes and mediates intracellular cholesterol trafficking via binding of cholesterol to its N-terminal domain. NPC1 transports low-density lipoproteins to late endosomal/lysosomal compartments where they are hydrolyzed and released as free cholesterol. NPC1, in addition to FTO, MC4R, and PTER has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NPC1
Gene Accession No. O15118
Gene Alias A430089E03Rik; C85354; Cdig2; D18Ertd139e; D18Ertd723e; FLJ98532; I79_014078; lcsd; Niemann Pick type C1; Niemann-Pick C disease protein; niemann-Pick C1 protein; Niemann-Pick disease, type C1; Niemann-Pick type C1; Niemann-Pick type C1 protein; nmf164; NPC; NPC intracellular cholesterol transporter 1; NPC1; sphingomyelinosis; spm; truncated Niemann-Pick C1
Gene Symbols NPC1
Host Species Rabbit
Immunogen Recombinant Human NPC intracellular cholesterol transporter 1 protein (23-269AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4864
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.