Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPC1L1 Polyclonal Antibody

Catalog No. PIPA5143865
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143865 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143865 Supplier Invitrogen™ Supplier No. PA5143865

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.

Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPC1L1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Npc1l1
Gene Accession No. Q6T3U3, Q6T3U4, Q9UHC9
Gene Alias 9130221N23Rik; BC 400-127; Gm243; Niemann-Pick C1-like protein 1; Niemann-Pick C3; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1 like intracellular cholesterol transporter 1; NPC11L1; NPC1L1; NPC1-like 1; NPC1-like intracellular cholesterol transporter 1; NPC3
Gene Symbols Npc1l1
Host Species Rabbit
Immunogen E.coli-derived human Niemann Pick C1 Like 1/NPC1L1 recombinant protein (Position: L778-A1231).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237636, 29881, 432367
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.