Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPC1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572938 Supplier Invitrogen™ Supplier No. PA572938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPC1L1 Polyclonal Antibody for Western Blot, IHC (P), IP

Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPC1L1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Npc1l1
Gene Accession No. Q6T3U3, Q6T3U4, Q9UHC9
Gene Alias 9130221N23Rik; BC 400-127; Gm243; Niemann-Pick C1-like protein 1; Niemann-Pick C3; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1 like intracellular cholesterol transporter 1; NPC11L1; NPC1L1; NPC1-like 1; NPC1-like intracellular cholesterol transporter 1; NPC3
Gene Symbols Npc1l1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 500-600).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237636, 29881, 432367
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.