Learn More
Invitrogen™ NPC1L1 Polyclonal Antibody

Description
The target sequence has 93% sequence homology with human.
Specifications
Specifications
| Antigen | NPC1L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate, tris citrate with 0.1% sodium azide; pH 7-8 |
| Gene | Npc1l1 |
| Gene Accession No. | Q6T3U3, Q6T3U4, Q9UHC9 |
| Gene Alias | 9130221N23Rik; BC 400-127; Gm243; Niemann-Pick C1-like protein 1; Niemann-Pick C3; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1 like intracellular cholesterol transporter 1; NPC11L1; NPC1L1; NPC1-like 1; NPC1-like intracellular cholesterol transporter 1; NPC3 |
| Gene Symbols | Npc1l1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.