Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPC2/Niemann Pick C2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11736R

Description
This gene encodes a protein containing a lipid recognition domain. The encoded protein may function in regulating the transport of cholesterol through the late endosomal/lysosomal system. Mutations in this gene have been associated with Niemann-Pick disease, type C2 and frontal lobe atrophy.Specifications
NPC2/Niemann Pick C2 | |
Polyclonal | |
Unconjugated | |
NPC2 | |
2700012J19Rik; AA408070; AU045843; EDDM1; epididymal protein 1; epididymal secretory protein 1; Epididymal secretory protein E1; HE1; Human epididymis-specific protein 1; mE1; MGC1333; Niemann Pick type C2; Niemann Pick type C2 protein homolog; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; Niemann-Pick type C2; NPC intracellular cholesterol transporter 2; Npc2; NP-C2; re1; tissue-specific secretory protein | |
Rabbit | |
Protein A | |
RUO | |
10577 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P61916 | |
NPC2 | |
KLH conjugated synthetic peptide derived from human Niemann Pick C2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction