Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579755
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579755 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579755 Supplier Invitrogen™ Supplier No. PA579755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SK-OV-3 whole cell. IHC: human lung cancer tissue, human intestinal cancer tissue.

This gene encodes a protein containing a lipid recognition domain. The encoded protein may function in regulating the transport of cholesterol through the late endosomal/lysosomal system. Mutations in this gene have been associated with Niemann-Pick disease, type C2 and frontal lobe atrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene NPC2
Gene Accession No. P61916
Gene Alias 2700012J19Rik; AA408070; AU045843; EDDM1; epididymal protein 1; epididymal secretory protein 1; epididymal secretory protein E1; HE1; human epididymis-specific protein 1; mE1; MGC1333; Niemann Pick type C2; niemann Pick type C2 protein homolog; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; Niemann-Pick type C2; NPC intracellular cholesterol transporter 2; Npc2; NP-C2; re1; tissue-specific secretory protein
Gene Symbols NPC2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human Niemann Pick C2 (KSEYPSIKLVVEWQLQDDKNQSLFCWEIPVQIVS).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10577
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.