Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPDC1 Monoclonal Antibody (16E4)
GREENER_CHOICE

Catalog No. 01674060
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-060 Supplier Invitrogen™ Supplier No. MA112777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control for this product is brain tissue lysate.

The discovery of NPDC-1 (neuronal proliferation, differentiation and control protein 1) was the result of a study of genes that were preferentially expressed in immortalized neural precursor cell lines at the onset of contact inhibition of proliferation and subsequent differentiation. It was further found that transfection of transformed cells with NPDC-1 led to increased doubling times and suppression of many characteristics of transformation. NPDC-1 is differentially expressed in brain and lung and has been shown to interact directly with the transcription factor E2F-1 as well as D-type cyclins and cdk2. NPDC-1 has been identified as a modulator of transcriptiona levents mediated by retinoic acid and the protein level and activity of NPDC-1 have been shown to be regulated by phosphorylation mediated proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPDC1
Applications Western Blot
Classification Monoclonal
Clone 16E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 3mM sodium azide; pH 7.5
Gene NPDC1
Gene Accession No. Q64322, Q9NQX5
Gene Alias AI314472; CAB; CAB-; CAB1; CAB-1; DKFZP586J0523; Neural proliferation differentiation and control protein 1; neural proliferation differentiation control-1 protein; neural proliferation, differentiation and control 1; neural proliferation, differentiation and control gene 1; neural proliferation, differentiation and control, 1; NPDC1; NPDC-1; RP23-47P18.6
Gene Symbols NPDC1
Host Species Mouse
Immunogen Synthetic peptide corresponding to human NPDC-1.
Purification Method Ion-exchange Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18146, 296562, 56654
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.