Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582338 Supplier Invitrogen™ Supplier No. PA582338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWCRLQREIR LTQKADYATA KAPGSPAAPR ISPGDQRLAQ SAEMYHYQHQ RQQMLCLERH KEPPKELDTA SSDEENEDGD FTVYECPGLA PTGEMEVRNP LFDHAALSAP LPAPSSPPAL P Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.

The discovery of NPDC-1 (neuronal proliferation, differentiation and control protein 1) was the result of a study of genes that were preferentially expressed in immortalized neural precursor cell lines at the onset of contact inhibition of proliferation and subsequent differentiation. It was further found that transfection of transformed cells with NPDC-1 led to increased doubling times and suppression of many characteristics of transformation. NPDC-1 is differentially expressed in brain and lung and has been shown to interact directly with the transcription factor E2F-1 as well as D-type cyclins and cdk2. NPDC-1 has been identified as a modulator of transcriptiona levents mediated by retinoic acid and the protein level and activity of NPDC-1 have been shown to be regulated by phosphorylation mediated proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPDC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPDC1
Gene Accession No. Q9NQX5
Gene Alias AI314472; CAB; CAB-; CAB1; CAB-1; DKFZP586J0523; Neural proliferation differentiation and control protein 1; neural proliferation differentiation control-1 protein; neural proliferation, differentiation and control 1; neural proliferation, differentiation and control gene 1; neural proliferation, differentiation and control, 1; NPDC1; NPDC-1; RP23-47P18.6
Gene Symbols NPDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPDC1. Recombinant protein control fragment (Product #RP-89897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.