Learn More
Invitrogen™ NPEPL1 Monoclonal Antibody (3F8-1A6)

Description
Peptide Sequence: MANVGLQFQA SAGDSDPQSR PLLLLGQLHH LHRVPWSHVR GKLQPRVTEE LWQAALSTLN PNPTDSCPLY LNYATVAALP CRVSRHNSPS AAHFITRLVR TCLPPGAHRC IVMVCEQPEV FASACALARA FPLFTHRSGA SRRLEKKTVT VEFFLVGQDN GPVEVSTLQC LANATDGVRL AARIVDTPCN EMNTDTFLEE INKVGKELGI IPTIIRDEEL KTRGFGGIYG VGKAALHPPA LAVLSHTPDG ATQTIAWVGK GIVYDTGGLS IKGKTTMPGM KRDCGGAAAV LGAFRAAIKQ GFKDNLHAVF CLAENSVGPN ATRPDDIHLL YSGKTVEINN TDAEGRLVLA DGVSYACKDL GADIILDMAT LTGAQGIATG KYHAAVLTNS AEWEAACVKA GRKCGDLVHP LVYCPELHFS EFTSAVADMK NSVADRDNSP SSCAGLFIAS HIGFDWPGVW VHLDIAAPVH AGERATGFGV ALLLALFGRA SEDPLLNLVS PLGCEVDVEE GDLGRDSKRR RLV.
Specifications
Specifications
| Antigen | NPEPL1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3F8-1A6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NPEPL1 |
| Gene Accession No. | Q8NDH3 |
| Gene Alias | Aminopeptidase-like 1; bA261P9.2; BC023239; C85514; KIAA1974; mKIAA1974; NPEPL1; Probable aminopeptidase NPEPL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.