Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPEPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583328
Description
Immunogen sequence: RAFPLFTHRS GASRRLEKKT VTVEFFLVGQ DNGPVEVSTL QCLANATDGV RLAARIVDTP CNEMNTDTFL EEINKVGKEL GIIPTIIRDE ELKTRGFG.
Probably catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.
Specifications
NPEPL1 | |
Polyclonal | |
Unconjugated | |
NPEPL1 | |
Aminopeptidase-like 1; bA261P9.2; BC023239; C85514; KIAA1974; mKIAA1974; NPEPL1; Probable aminopeptidase NPEPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79716 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDH3 | |
NPEPL1 | |
Recombinant protein corresponding to Human NPEPL1. Recombinant protein control fragment (Product #RP-95876). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction