Learn More
NPEPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 172111AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-1443
Only null left
Add to Cart
Description
Description
Probably catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.
Specifications
Specifications
NPEPL1
Polyclonal
Unconjugated
NPEPL1
aminopeptidase like 1, KIAA1974, NPEPL1, Probable aminopeptidase NPEPL1
Rabbit
Antigen Affinity Chromatography
RUO
228961, 311671, 79716
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6NSR8, Q8NDH3
Npepl1
NPEPL1 Fusion Protein Ag10994
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.