Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPEPPS Polyclonal Antibody

Catalog No. PIPA5121735
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121735 20 μL
PIPA5119383 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121735 Supplier Invitrogen™ Supplier No. PA5121735
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPEPPS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes the puromycin-sensitive aminopeptidase, a zinc metallopeptidase which hydrolyzes amino acids from the N-terminus of its substrate. The protein has been localized to both the cytoplasm and to cellular membranes. This enzyme degrades enkaphalins in the brain, and studies in mouse suggest that it is involved in proteolytic events regulating the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPEPPS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NPEPPS
Gene Accession No. P55786
Gene Alias AAP-S; aminopeptidase puromycin sensitive; cytosol alanyl aminopeptidase; goku; metalloproteinase MP100; MP100; NPEPPS; Psa; Puromycin-sensitive aminopeptidase; R74825
Gene Symbols NPEPPS
Host Species Rabbit
Immunogen Recombinant protein Puromycin Sensitive Aminopeptidase. The antigen corresponds to amino acid range Gly584-Gln793 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414413, 9520
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.