Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPEPPS/PSA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981582
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1582 Supplier Bioss Supplier No. BS11684R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes the puromycin-sensitive aminopeptidase, a zinc metallopeptidase which hydrolyzes amino acids from the N-terminus of its substrate. The protein has been localized to both the cytoplasm and to cellular membranes. This enzyme degrades enkaphalins in the brain, and studies in mouse suggest that it is involved in proteolytic events regulating the cell cycle.

Specifications

Antigen NPEPPS/PSA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPEPPS
Gene Accession No. P55786
Gene Alias AAP-S; aminopeptidase puromycin sensitive; Cytosol alanyl aminopeptidase; goku; metalloproteinase MP100; MP100; Npepps; PSA; Puromycin-sensitive aminopeptidase; R74825
Gene Symbols NPEPPS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NPEPPS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9520
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.