Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPFF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596547
Description
Immunogen sequence: AEEDSEPLPP QDAQTSGSLL HYLLQAMERP GRSQAFLFQP QRFGRNTQGS WRNEWLSPRA GEGLNSQFWS LAAPQRFGKK; Positive Samples: Rat plasma.
FMFRamide-related protein precursor plays a role in the regulation of heart rate and blood pressure and the modulation of morphine-induced antinociception. FMRFAL encodes a preproprotein which is cleaved to form two active peptides with similar function.
Specifications
NPFF | |
Polyclonal | |
Unconjugated | |
NPFF | |
FMRFAL; FMRFamide-related peptides; Neuropeptide AF; Neuropeptide AF-like; Neuropeptide FF; neuropeptide FF-amide peptide precursor; Neuropeptide SF; NPAF; Npff; NPSF; Pro-FMRFamide-related neuropeptide FF | |
Rabbit | |
Affinity chromatography | |
RUO | |
60337, 8620 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15130, Q9WVA9 | |
NPFF | |
Recombinant fusion protein containing a sequence corresponding to amino acids 34-113 of human NPFF (NP_0037081). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction