Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPFF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115244 Supplier Thermo Scientific Supplier No. PA5115244
Add to Cart
Add to Cart

NPFF Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPFF
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NPFF
Gene Accession No. O15130
Gene Alias FMRFAL; FMRFamide-related peptides; Neuropeptide AF; Neuropeptide AF-like; Neuropeptide FF; neuropeptide FF-amide peptide precursor; Neuropeptide SF; NPAF; Npff; NPSF; Pro-FMRFamide-related neuropeptide FF
Gene Symbols NPFF
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPFF(Accession O15130), corresponding to amino acid residues Q58-Q108.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8620
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.