Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPFF Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8150R

Description
FMFRamide-related protein precursor plays a role in the regulation of heart rate and blood pressure and the modulation of morphine-induced antinociception. FMRFAL encodes a preproprotein which is cleaved to form two active peptides with similar function.Specifications
NPFF | |
Polyclonal | |
Unconjugated | |
NPFF | |
FMRFAL; FMRFamide-related peptides; Neuropeptide AF; Neuropeptide AF-like; Neuropeptide FF; neuropeptide FF-amide peptide precursor; Neuropeptide SF; NPAF; NPFF; NPSF; Pro-FMRFamide-related neuropeptide FF | |
Rabbit | |
Protein A | |
RUO | |
8620 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O15130 | |
NPFF | |
KLH conjugated synthetic peptide derived from human NPFF/FMRFamide. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction