Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPFF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518891
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518891 Supplier Invitrogen™ Supplier No. PA518891
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The central nervous system neuropeptide FF (NPFF), is believed to play a role in pain modulation and opiate tolerance as well as other physiological functions. Two G protein-coupled receptors, NPFF1 and NPFF2, were isolated from the human central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPFF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NPFFR1
Gene Accession No. Q9GZQ6
Gene Alias ff1; FLJ10751; G protein-coupled receptor 147; Gm236; Gpr147; G-protein coupled receptor 147; G-protein coupled receptor NPFF1R; neuropeptide FF 1; neuropeptide FF receptor 1; NPFF1; NPFF1 receptor; NPFF1R1; NPFFR1; NPFF-R1; OT7T022; RFamide-related peptide receptor; RFamide-related peptide receptor OT7T022
Gene Symbols NPFFR1
Host Species Goat
Immunogen Synthetic peptide sequence (RPSGSHKEAYSERP) corresponding to the internal amino acids of NPFFR1 (aa 351-365).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64106
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.