Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPFF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111081 Supplier Invitrogen™ Supplier No. PA5111081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIGRKLCQAP GPAPGGEEAA DPRASRRR Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 70%.

The central nervous system neuropeptide FF (NPFF), is believed to play a role in pain modulation and opiate tolerance as well as other physiological functions. Two G protein-coupled receptors, NPFF1 and NPFF2, were isolated from the human central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPFF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPFFR1
Gene Accession No. Q9GZQ6
Gene Alias ff1; FLJ10751; G protein-coupled receptor 147; Gm236; Gpr147; G-protein coupled receptor 147; G-protein coupled receptor NPFF1R; neuropeptide FF 1; neuropeptide FF receptor 1; NPFF1; NPFF1 receptor; NPFF1R1; NPFFR1; NPFF-R1; OT7T022; RFamide-related peptide receptor; RFamide-related peptide receptor OT7T022
Gene Symbols NPFFR1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human NPFF1. Recombinant protein control fragment (Product #RP-95313).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.