Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPFF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588020 Supplier Invitrogen™ Supplier No. PA588020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNSFFGTPAA SWCLLESDVS SAPDKEAGRE RRALSVQQRG GPAWSGSLEW SRQSAGDRRR LGLSRQTAKS SWSRSRDRTC CCRRAWWILV PAADRARRER FIMNEKWDTN SSENWHPIWN VNDTKHHLYS DINITYVNYY; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein.

Receptor for NPAF (A-18-F-amide) and NPFF (F-8-F-amide) neuropeptides, also known as morphine-modulating peptides. Can also be activated by a variety of naturally occurring or synthetic FMRF-amide like ligands. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPFF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NPFFR2
Gene Accession No. Q924H0, Q9EQD2
Gene Alias G protein-coupled receptor 74; Gpr74; G-protein coupled receptor 74; G-protein coupled receptor HLWAR77; HG31; HLWAR77; neuropeptide FF 2; neuropeptide FF receptor 2; Neuropeptide G-protein coupled receptor; neuropeptide NPFF receptor; Npff2; NPFF2 receptor; Npffr2; NPFF-R2; NPGP receptor; Npgpr
Gene Symbols NPFFR2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human NPFFR2 (NP_0048762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104443, 78964
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.