Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPFF2 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278148
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8148 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8148 Supplier Leinco Technologies Inc Supplier No. 56111100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPFF2 Polyclonal antibody specifically detects NPFF2 in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen NPFF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NPFFR2
Gene Accession No. Q9Y5X5
Gene Alias G protein-coupled receptor 74; Gpr74; G-protein coupled receptor 74; G-protein coupled receptor HLWAR77; HG31; HLWAR77; neuropeptide FF 2; neuropeptide FF receptor 2; Neuropeptide G-protein coupled receptor; neuropeptide NPFF receptor; Npff2; NPFF2 receptor; Npffr2; NPFF-R2; NPGP receptor; Npgpr
Host Species Rabbit
Immunogen N-terminal sequence MNEKWDTNSSENWHPI (4-19) and C-terminal sequence (506-522) ELVMEELKETTNSSEI of human NPFF2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10886
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.