Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHP1 Mouse anti-Human, Unconjugated, Clone: 1M4, MilliporeSigma™

Catalog No. MABS21852MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21852MI 25 μL
MABS21851MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21852MI Supplier MilliporeSigma Supplier No. MABS218525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPHP1 Monoclonal antibody specifically detects NPHP1 in Human samples. It is validated for Western Blotting, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPHP1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1M4
Conjugate Unconjugated
Gene Accession No. NP_997064.2
Gene Alias Nephrocystin-1;Juvenile nephronophthisis 1 protein
Gene Symbols NPHP1, NPH1
Host Species Mouse
Immunogen A synthetic polypeptide fused by 16 different NPHP1 peptide sequences predicted to have the highest antigenicity. The sequence was converted to cDNA and the polypeptide was expressed and purified from E. coli.
Molecular Weight of Antigen ∼92 kDa observed; 83.30 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 1M4 is a mouse monoclonal antibody that specifically detects human Nephrocystin-1 (NPHP1).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.