Learn More
Invitrogen™ NPHP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593140
Description
Immunogen sequence: PDLGILFELG ISYIRNSTGE RGELSCGWVF LKLFDASGVP IPAKTYELFL NGGTPYEKGI EVDPSISRRA HGSVFYQIMT MRRQPQLLVK LRSLNRRSRN VLSLLPETLI GNMCSIHLLI FYRQILGDVL LKDRMSLQST DLISHPMLAT FPMLLEQPDV MDALRSSWAG KESTLKRSEK RDKEFLKSTF LLVYHDCVLP LLHSTRLPPF RWAEEETETA RWKVITDFLK QNQENQGALQ ALLSPDGVHE PFDLSEQTYD FLGEMRKNAV; Positive Samples: Mouse lung, Rat brain; Cellular Location: Cell junction, Cell projection, Cytoplasm, adherens junction, cilium, cilium axoneme, cytoskeleton, tight junction.
Juvenile nephronophthisis type 1 is caused by mutations of NPHP1, the gene encoding for nephrocystin. Nephrocystin interacts with p130Cas (BCAR1), proline-rich tyrosine kinase-2 (PTK2B or Pyk2), and tensin in embryonic kidney and testis, indicating that these proteins participate in a common signaling pathway. Nephrocystin and p130Cas interact in mammalian cells and both proteins prominently localize at or near sites of cell-cell contact in polarized Madin-Darby canine kidney epithelial cells. Expression of nephrocystin results in phosphorylation of Pyk2 on Tyrosine 402 as well as activation of downstream mitogen-activated protein kinases, such as ERK1 and ERK2. Nephrocystin contains a src-homology 3 (SH3) domain, which is highly conserved throughout evolution.
Specifications
NPHP1 | |
Polyclonal | |
Unconjugated | |
NPHP1 | |
JBTS4; juvenile nephronophthisis 1 protein; nephrocystin 1; Nephrocystin1; nephrocystin-1; nephronophthisis 1 (juvenile); nephronophthisis 1 (juvenile) homolog; nephronophthisis 1 (juvenile) homolog (human); NPH1; NPHP1; SLSN1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
296136, 4867, 53885 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.97 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15259, Q9QY53 | |
NPHP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 463-732 of human NPHP1 (NP_9970642). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.