Learn More
Invitrogen™ NPHP3 Monoclonal Antibody (3B1)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524451
Catalog No. PIMA524451
Only null left
Add to Cart
Description
Description
Peptide Sequence: NQELLSMGRR EAKLDTENKR LRAELQALQK TYQKILREKE SALEAKYQAM ERAATFEHDR DKVKRQFKIF RETKENEIQD LLRAKRELES KLQRLQAQGI.
Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPHP3
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Q7Z494
NPHP3
NPHP3 (NP_694972.3, 106 a.a. ~ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
3B1
Unconjugated
NPHP3
3632410F03Rik; AI550417; C230078J01; CFAP31; cilia and flagella associated protein 31; D330020E01Rik; Kiaa2000; Meckel syndrome, type 7; MKS7; Nephrocystin-3; nephronophthisis 3 (adolescent); NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3
Mouse
Protein A
RUO
27031
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.