Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPHP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552656 Supplier Invitrogen™ Supplier No. PA552656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEFAHSSIDV EGPFANVNRD DWDIAVASLL QVTPLFSHSL WSNTVRCYLI YTDETQPEMD LFLKDYSPKL KRMCETMGYF FHAVYFPIDV ENQYLTVRKW EIEKS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.

Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPHP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPHP3
Gene Accession No. Q7Z494
Gene Alias 3632410F03Rik; AI550417; C230078J01; CFAP31; cilia and flagella associated protein 31; D330020E01Rik; Kiaa2000; Meckel syndrome, type 7; MKS7; Nephrocystin-3; nephronophthisis 3 (adolescent); NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3
Gene Symbols NPHP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPHP3. Recombinant protein control fragment (Product #RP-89775).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27031
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.