Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPHP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113061
Description
NPHP3 Polyclonal Antibody for Western Blot, ELISA
Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
Specifications
NPHP3 | |
Polyclonal | |
Unconjugated | |
NPHP3 | |
3632410F03Rik; AI550417; C230078J01; CFAP31; cilia and flagella associated protein 31; D330020E01Rik; Kiaa2000; Meckel syndrome, type 7; MKS7; Nephrocystin-3; nephronophthisis 3 (adolescent); NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27031, 74025 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TNH6, Q7Z494 | |
NPHP3 | |
Recombinant Human Nephrocystin-3 protein (1-130AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction