Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPHP3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA597123

Catalog No. PIPA597123


Only null left
Add to Cart

Description

Description

Immunogen sequence: MGTASSLVSP AGGEVIEDTY GAGGGEACEI PVEVKPKARL LRNSFRRGAG AAAGAGPGSL PRGVGAGGLL GASFKSTGSS VPELEYAAAE YERLRKEYEI FRVSKNQELL SMGRREAKLD TENKRLRAEL; Positive Samples: 293T, Mouse liver; Cellular Location: Cell projection, cilium.

Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPHP3
Polyclonal
Unconjugated
NPHP3
3632410F03Rik; AI550417; C230078J01; CFAP31; cilia and flagella associated protein 31; D330020E01Rik; Kiaa2000; Meckel syndrome, type 7; MKS7; Nephrocystin-3; nephronophthisis 3 (adolescent); NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3
Rabbit
Affinity Chromatography
RUO
27031, 74025
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TNH6, Q7Z494
NPHP3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human NPHP3 (NP_6949723).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.