Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPHP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139863 Supplier Invitrogen™ Supplier No. PA5139863
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FTQNVLVPPH PQRARQPWKE STAFQCVLKW LDGPVIRQGV LEVLSEVECH LRVSFFDVTY RHFFGRTWKT TVKPTKRPPS RIVFNEPLYF HTSLNHPHIV AVVEVVAEGK KRDGS Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 78%.

This gene encodes a protein involved in renal tubular development and function. This protein interacts with nephrocystin, and belongs to a multifunctional complex that is localized to actin- and microtubule-based structures. Mutations in this gene are associated with nephronophthisis type 4, a renal disease, and with Senior-Loken syndrome type 4, a combination of nephronophthisis and retinitis pigmentosa. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPHP4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPHP4
Gene Accession No. O75161
Gene Alias 4930564O18Rik; KIAA0673; nephrocystin 4; nephrocystin-4; nephronophthisis 4; nephronophthisis 4 (juvenile) homolog (human); nephroretinin; nmf192; NPHP4; POC10; POC10 centriolar protein homolog; SLSN4
Gene Symbols NPHP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPHP4. Recombinant protein control fragment (Product #RP-109050).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.