Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHS1, Rabbit anti-Human, Clone: G17-H, Abnova™

Catalog No. 89998746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-746 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-746 Supplier Abnova Corporation Supplier No. MAB15906
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen NPHS1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G17-H
Conjugate Unconjugated
Dilution ELISA (1:20000-1:50000) Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide).
Gene nephrosis 1, congenital, Finnish type (nephrin)
Gene Alias CNF/NPHN
Gene Symbols NPHS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 53-101 at C-terminus of the first Ig-like domain of human NPHS1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4868
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.