Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732859 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2859 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2859 Supplier Proteintech Group Inc Supplier No. 203841AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.

Specifications

Antigen NPHS2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NPHS2
Gene Accession No. Q8K4G9, Q91X05, Q9NP85
Gene Alias NPHS2, PDCN, Podocin, SRN1
Gene Symbols Nphs2
Host Species Rabbit
Immunogen NPHS2 Fusion Protein Ag14213
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170484, 170672, 7827
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.