Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPLOC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100647 Supplier Invitrogen™ Supplier No. PA5100647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NPLOC4.

NPLOC4 gene ontology annotations related to this gene include ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPLOC4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NPLOC4
Gene Accession No. P60670, Q8TAT6, Q9ES54
Gene Alias AK129375; FLJ20657; homolog of yeast nuclear protein localization 4; KIAA1499; mKIAA1499; NPL4; NPL4 homolog, ubiquitin recognition factor; Nploc4; NPLOC4 ubiquitin recognition factor; nuclear protein localization 4 homolog; nuclear protein localization 4 homolog (S. cerevisiae); nuclear protein localization protein 4 homolog; Protein NPL4
Gene Symbols NPLOC4
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPLOC4(Accession Q8TAT6), corresponding to amino acid residues G173-V223.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140639, 217365, 55666
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.