Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPLOC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554450
Description
Immunogen sequence: VRDECLLPCK DAPELGYAKE SSSEQYVPDV FYKDVDKFGN EITQLARPLP VEYLIIDITT TFPKDPVYTF SISQNPFPIE NRDVLGETQD FHSLATYLSQ NTSSVFLDTI SDFHLLLFLV TNE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
NPLOC4 gene ontology annotations related to this gene include ubiquitin-dependent protein catabolic process.
Specifications
NPLOC4 | |
Polyclonal | |
Unconjugated | |
NPLOC4 | |
AK129375; FLJ20657; homolog of yeast nuclear protein localization 4; KIAA1499; mKIAA1499; NPL4; NPL4 homolog, ubiquitin recognition factor; Nploc4; NPLOC4 ubiquitin recognition factor; nuclear protein localization 4 homolog; nuclear protein localization 4 homolog (S. cerevisiae); nuclear protein localization protein 4 homolog; Protein NPL4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140639, 217365, 55666 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P60670, Q8TAT6, Q9ES54 | |
NPLOC4 | |
Recombinant protein corresponding to Human NPLOC4. Recombinant protein control fragment (Product #RP-93491). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction