Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPM1 Antibody (NPM1/3285), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308745AF532
Description
NPM1 Monoclonal antibody specifically detects NPM1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
NPM1 | |
Monoclonal | |
Alexa Fluor 532 | |
B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin | |
Recombinant human NPM1 protein fragment (aa185-287) (exact sequence is proprietary) (Uniprot: P06748) | |
100 μg | |
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Signal Transduction, Tumor Suppressors | |
4869 | |
Store at 4°C in the dark. | |
IgG2c κ |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
NPM1/3285 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction