Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPM1 Mouse anti-Human, Clone: 1042CT5.2.1, Abnova™

Catalog No. 89934706 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-706 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-706 Supplier Abnova Corporation Supplier No. MAB12333
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human NPM1.

NPM1 is a ubiquitously expressed nucleolar protein that shuttles between the nucleus and cytoplasm. It is implicated in multiple functions, including ribosomal protein assembly and transport, control of centrosome duplication, and regulation of the tumor suppressor ARF (MIM 600160). NPM1 mutations that relocalize NPM1 from the nucleus into the cytoplasm are associated with development of acute myeloid leukemia (AML; MIM 601626) (Garzon et al., 2008 [PubMed 18308931]).[supplied by OMIM]

Specifications

Antigen NPM1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1042CT5.2.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human NPM1.
Dilution Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias B23/MGC104254/NPM
Gene Symbols NPM1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human NPM1.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4869
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.