Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPM1 Antibody (NPM1/1902) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279874
Description
NPM1 Monoclonal specifically detects NPM1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
NPM1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
NPM1/1902 | |
Unconjugated | |
10 mM PBS with No Preservative | |
NPM1 | |
Recombinant fragment (around aa185-287) of human NPM1 protein (exact sequence is proprietary) (Uniprot: P06748 ) | |
100 μg | |
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Phospho Specific, Signal Transduction, Tumor Suppressors | |
Recognizes a 33kDa glycoprotein, identified as NPM1 (NPM). It is predominantly localized in the nucleus of cells in most tissues. NPM is involved in ribosomal assembly and rRNA transport. It is an abundant protein that is highly phosphorylated by Cdc2 kinase during mitosis. This phosphoprotein moves between the nucleus and the cytoplasm. It is thought to be involved in several processes including regulation of the ARF/p53 pathway. A number of genes are fusion partners, in particular the anaplastic lymphoma kinase gene on chromosome 2. Mutations in exon 12 affecting the C-terminus of the protein are associated with an aberrant cytoplasmic location. Mutations in this gene are associated with acute myeloid leukemia. The antibody may be a useful aid for classification of acute myeloid leukemia. | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction