Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPM1, p Thr199 Antibody (SR1550), Novus Biologicals™
SDP

Catalog No. p-200075851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387963 100 μg
NB387962 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387963 Supplier Novus Biologicals Supplier No. NBP321600100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NPM1 Monoclonal antibody specifically detects NPM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NPM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1550
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-NPM1 [Thr199] (Uniprot #: P06748)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Phospho Specific, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4869
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.