Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525254
Description
NPM3 Polyclonal Antibody for Western Blot, Flow
The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
Specifications
NPM3 | |
Polyclonal | |
Unconjugated | |
NPM3 | |
Npm3; Nub1; nucleophosmin/nucleoplasmin 3; nucleophosmin/nucleoplasmin family, member 3; nucleophosmin/nucleoplasmin, 3; nucleoplasmin 3; nucleoplasmin-3; PORMIN; RGD1565542; TMEM123 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10360, 18150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O75607, Q9CPP0 | |
NPM3 | |
KLH conjugated synthetic peptide between 30-58 amino acids from the N-terminal region of human NPM3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction