Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116760 Supplier Invitrogen™ Supplier No. PA5116760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Raji Immunogen sequence: APVTMDSFFF GCELSGHTRS FTFKVEEEDD AEHVLALTML CLTEGAKDEC NVVEVVARNH DHQEIAVPVA NLKLSCQPML SLDDFQLQPP VTFRLKSGSG PVRITGRHQI V.

The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPM3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NPM3
Gene Accession No. O75607
Gene Alias Npm3; Nub1; nucleophosmin/nucleoplasmin 3; nucleophosmin/nucleoplasmin family, member 3; nucleophosmin/nucleoplasmin, 3; nucleoplasmin 3; nucleoplasmin-3; PORMIN; RGD1565542; TMEM123
Gene Symbols NPM3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human NPM3 (NP_0089241).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10360
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.